siRNA Details
Gene Symbol: FLVCR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027285.1 | 1 | 318 | |||
NR_027286.1 | 1 | 318 | ||||
GenBank mRNA | AY030238.1 | - | 1 | 257 | ||
AY030239.1 | - | 1 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
