siRNA Details
Gene Symbol: FAM180B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164379.1 | NP_001157851.1 | 3 | 1187 | ||
GenBank mRNA | BC065704.1 | - | 3 | 1176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
