siRNA Details
Gene Symbol: NFE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085386.1 | NP_001078855.1 | 1 | 152 | ||
GenBank mRNA | AY258907.1 | - | 1 | 152 | ||
BC036938.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
