siRNA Details
Gene Symbol: RPL27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000990.4 | NP_000981.1 | 5 | 4272 | ||
GenBank mRNA | AK125453.1 | - | 1 | 2222 | ||
BC004284.1 | - | 1 | 1584 | |||
BC014919.1 | - | 1 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
