siRNA Details
Gene Symbol: ERVH48-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308491.1 | NP_001295420.1 | 2 | 444 | ||
GenBank mRNA | AB610407.1 | - | 2 | 444 | ||
BC005107.1 | - | 2 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
