siRNA Details
Gene Symbol: CCDC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025055.4 | NP_079331.3 | 3 | 629 | ||
XM_005254692.1 | XP_005254749.1 | 3 | 629 | |||
XM_011522085.2 | XP_011520387.1 | 4 | 1474 | |||
XM_011522086.1 | XP_011520388.1 | 3 | 322 | |||
XM_011522087.2 | XP_011520389.1 | 4 | 460 | |||
XM_011522088.1 | XP_011520390.1 | 3 | 330 | |||
XM_017022623.1 | XP_016878112.1 | 4 | 1475 | |||
XM_017022624.1 | XP_016878113.1 | 4 | 1474 | |||
XM_017022625.1 | XP_016878114.1 | 4 | 1474 | |||
XM_017022626.1 | XP_016878115.1 | 4 | 1474 | |||
XM_017022627.1 | XP_016878116.1 | 4 | 1474 | |||
XM_017022628.1 | XP_016878117.1 | 4 | 1474 | |||
XM_017022629.1 | XP_016878118.1 | 6 | 1215 | |||
XM_017022630.1 | XP_016878119.1 | 4 | 1476 | |||
XM_017022631.1 | XP_016878120.1 | 4 | 1475 | |||
XM_017022633.1 | XP_016878122.1 | 4 | 1484 | |||
GenBank mRNA | AK057417.1 | - | 3 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
