siRNA Details
Gene Symbol: MYBPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010985.2 | NP_001010985.2 | 7 | 1093 | ||
NM_001265613.1 | NP_001252542.1 | 7 | 1024 | |||
XM_017001173.1 | XP_016856662.1 | 7 | 3446 | |||
XM_017001174.1 | XP_016856663.1 | 7 | 3446 | |||
XM_017001175.1 | XP_016856664.1 | 7 | 3377 | |||
GenBank mRNA | AK129834.1 | - | 7 | 1093 | ||
BC108664.1 | - | 2 | 326 | |||
BC132900.1 | - | 7 | 1070 | 40146922 | ||
BC132902.1 | - | 7 | 1070 | 40146924 | ||
BC144471.1 | - | 7 | 1001 | 9052998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
