siRNA Details
Gene Symbol: RICTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285439.1 | NP_001272368.1 | 31 | 3634 | ||
NM_001285440.1 | NP_001272369.1 | 31 | 3632 | |||
NM_152756.4 | NP_689969.2 | 31 | 3634 | |||
XM_006714463.3 | XP_006714526.1 | 31 | 3647 | |||
XM_011514005.2 | XP_011512307.1 | 31 | 3647 | |||
XM_011514006.2 | XP_011512308.1 | 30 | 3451 | |||
XM_017009311.1 | XP_016864800.1 | 30 | 3567 | |||
XM_017009312.1 | XP_016864801.1 | 30 | 3567 | |||
XM_017009313.1 | XP_016864802.1 | 30 | 3588 | |||
XM_017009314.1 | XP_016864803.1 | 30 | 3557 | |||
XM_017009315.1 | XP_016864804.1 | 30 | 3557 | |||
GenBank mRNA | AB082530.1 | - | 16 | 2305 | ||
AL834497.1 | - | 3 | 812 | |||
AY515854.1 | - | 31 | 3630 | |||
BC051729.1 | - | 11 | 1632 | |||
BC137163.1 | - | 31 | 3627 | 9021160 | ||
BC137164.1 | - | 31 | 3627 | 9021161 | ||
BC144509.1 | - | 31 | 3627 | 9053036 | ||
CR749280.1 | - | 31 | 3613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
