siRNA Details
Gene Symbol: ZSCAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005249296.4 | XP_005249353.1 | 1 | 2330 | ||
XM_011514807.2 | XP_011513109.1 | 1 | 2330 | |||
XM_011514808.2 | XP_011513110.1 | 1 | 2330 | |||
XM_017011196.1 | XP_016866685.1 | 1 | 2330 | |||
GenBank mRNA | AK123280.1 | - | 1 | 399 |
Gene Symbol: LOC439938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095259.1 | - | 1 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
