siRNA Details
Gene Symbol: HRNR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009931.2 | NP_001009931.1 | 3 | 2307 | ||
GenBank mRNA | AB104446.1 | - | 1 | 80 | ||
AY396741.1 | - | 3 | 2307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
