siRNA Details
Gene Symbol: MRPS31P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002816.2 | 7, 8 | 1787 | |||
NR_051963.1 | 7, 8 | 1072 | ||||
NR_051964.1 | 5, 6 | 657 | ||||
GenBank mRNA | AK027536.1 | - | 7, 8 | 1787 | ||
AK307345.1 | - | 5, 6 | 610 | |||
AK309380.1 | - | 7, 8 | 1045 | |||
BX648914.1 | - | 7, 8 | 1062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
