siRNA Details
Gene Symbol: MRPS31P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002816.2 | 4 | 1414 | |||
NR_051963.1 | 4 | 699 | ||||
NR_051965.1 | 3 | 423 | ||||
GenBank mRNA | AK027536.1 | - | 4 | 1414 | ||
AK307124.1 | - | 3 | 379 | |||
AK309380.1 | - | 4 | 672 | |||
BX648914.1 | - | 4 | 689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
