siRNA Details
Gene Symbol: MRPS31P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002816.2 | 9 | 2166 | |||
NR_051963.1 | 9 | 1451 | ||||
NR_051964.1 | 7 | 1036 | ||||
NR_051965.1 | 8 | 1132 | ||||
GenBank mRNA | AK027536.1 | - | 9 | 2166 | ||
AK307345.1 | - | 7 | 989 | |||
BX648914.1 | - | 9 | 1441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
