siRNA Details
Gene Symbol: LOC100134317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029389.1 | 1 | 288 |
Gene Symbol: LOC284412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029390.1 | 1 | 2035 | |||
GenBank mRNA | AF289566.1 | - | 1 | 1274 | ||
BC045185.1 | - | 1 | 2042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
