siRNA Details
Gene Symbol: MBOAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080480.2 | NP_001073949.1 | 7 | 819 | ||
NR_073465.1 | 4 | 539 | ||||
XM_006714999.2 | XP_006715062.1 | 7 | 556 | |||
XM_006715000.3 | XP_006715063.1 | 7 | 757 | |||
XM_011514313.2 | XP_011512615.1 | 7 | 757 | |||
GenBank mRNA | AB305043.1 | - | 7 | 589 | ||
AK093994.1 | - | 1 | 18 | |||
AK131269.1 | - | 7 | 782 | |||
AK296857.1 | - | 4 | 539 | |||
AK304748.1 | - | 7 | 764 | |||
BC150650.1 | - | 7 | 765 | 9057021 | ||
BC150652.1 | - | 7 | 765 | 9057023 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
