siRNA Details
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518300.2 | XP_011516602.1 | 6 | 11095 | ||
XM_017014337.1 | XP_016869826.1 | 7 | 10068 | |||
XM_017014338.1 | XP_016869827.1 | 7 | 9954 | |||
GenBank mRNA | AK054857.1 | - | 1 | 1737 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
