siRNA Details
Gene Symbol: YAE1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282446.1 | NP_001269375.1 | 3 | 1615 | ||
GenBank mRNA | AK096857.1 | - | 1 | 1451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
