siRNA Details
Gene Symbol: LCN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178469.3 | NP_848564.2 | 3 | 556 | ||
XM_017014272.1 | XP_016869761.1 | 3 | 284 | |||
GenBank mRNA | AK090647.1 | - | 2 | 553 | ||
AK126902.1 | - | 2 | 1294 | |||
AY301268.1 | - | 3 | 276 | |||
BC130465.1 | - | 3 | 511 | 40146496 | ||
BC132714.1 | - | 3 | 511 | 40146736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
