siRNA Details
Gene Symbol: C19orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136482.1 | NP_001129954.1 | 2 | 230 | ||
XM_005259846.4 | XP_005259903.1 | 3 | 576 | |||
XM_005259847.4 | XP_005259904.1 | 3 | 418 | |||
GenBank mRNA | BC157863.1 | - | 2 | 230 | 9057082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
