siRNA Details
Gene Symbol: ZNF799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080821.2 | NP_001074290.1 | 4 | 1195 | ||
NM_001322497.1 | NP_001309426.1 | 4 | 1645 | |||
NM_001322498.1 | NP_001309427.1 | 5 | 1313 | |||
XM_017027452.1 | XP_016882941.1 | 3 | 988 | |||
GenBank mRNA | AK027660.1 | - | 4 | 1645 | ||
AL832890.1 | - | 3 | 2685 | |||
BC009517.1 | - | 1 | 165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
