siRNA Details
Gene Symbol: SLFN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129820.1 | NP_001123292.1 | 1 | 430 | ||
XM_017024576.1 | XP_016880065.1 | 3 | 599 | |||
XM_017024577.1 | XP_016880066.1 | 3 | 621 | |||
XM_017024578.1 | XP_016880067.1 | 2 | 488 | |||
XM_017024579.1 | XP_016880068.1 | 2 | 521 | |||
GenBank mRNA | BC140847.1 | - | 1 | 427 | 9021715 | |
BC157877.1 | - | 1 | 431 | 9057094 | ||
BC157879.1 | - | 1 | 438 | 9057095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
