siRNA Details
Gene Symbol: C17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113434.4 | NP_001106905.1 | 2 | 6842 | ||
GenBank mRNA | AK096256.1 | - | 1 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
