siRNA Details
Gene Symbol: MUC5AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304359.1 | NP_001291288.1 | 16633 | |||
GenBank mRNA | AJ001402.1 | - | 17, 18 | 2867 | ||
AL833060.1 | - | 11, 12 | 1009 | |||
BC033831.1 | - | 12, 13 | 1347 | |||
U06711.1 | - | 17, 18 | 2742 | |||
Z48314.1 | - | 17, 18 | 2476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
