siRNA Details
Gene Symbol: KIAA1671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145206.1 | NP_001138678.1 | 11 | 6744 | ||
XM_005261793.2 | XP_005261850.1 | 13 | 7057 | |||
XM_005261794.4 | XP_005261851.1 | 13 | 7010 | |||
XM_006724346.2 | XP_006724409.1 | 14 | 6996 | |||
GenBank mRNA | AB051458.1 | - | 8 | 2392 | ||
BC171801.1 | - | 12 | 6744 | 9054455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
