siRNA Details
Gene Symbol: FLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014342.2 | NP_001014364.1 | 3 | 3364 | ||
XM_011509531.2 | XP_011507833.1 | 4 | 2930 | |||
GenBank mRNA | AY827490.1 | - | 3 | 3365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
