siRNA Details
Gene Symbol: COL6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102608.1 | NP_001096078.1 | 7 | 3057 | ||
XM_005247121.4 | XP_005247178.1 | 9 | 3216 | |||
XM_011512423.2 | XP_011510725.1 | 8 | 3281 | |||
XM_011512424.2 | XP_011510726.1 | 8 | 3281 | |||
XM_011512425.2 | XP_011510727.1 | 10 | 3396 | |||
XM_011512426.2 | XP_011510728.1 | 10 | 3290 | |||
XM_011512428.2 | XP_011510730.1 | 9 | 3317 | |||
XM_011512431.2 | XP_011510733.1 | 8 | 3282 | |||
XM_011512432.2 | XP_011510734.1 | 8 | 3282 | |||
XM_017005711.1 | XP_016861200.1 | 11 | 3501 | |||
XM_017005712.1 | XP_016861201.1 | 10 | 3605 | |||
XM_017005713.1 | XP_016861202.1 | 11 | 3552 | |||
XM_017005714.1 | XP_016861203.1 | 9 | 9657 | |||
XM_017005715.1 | XP_016861204.1 | 10 | 3422 | |||
XM_017005716.1 | XP_016861205.1 | 8 | 3282 | |||
GenBank mRNA | AM774226.1 | - | 3 | 757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
