siRNA Details
Gene Symbol: COL6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102608.1 | NP_001096078.1 | 3 | 1058 | ||
XM_005247121.4 | XP_005247178.1 | 5 | 1217 | |||
XM_011512423.2 | XP_011510725.1 | 4 | 1282 | |||
XM_011512424.2 | XP_011510726.1 | 4 | 1282 | |||
XM_011512425.2 | XP_011510727.1 | 6 | 1397 | |||
XM_011512426.2 | XP_011510728.1 | 6 | 1291 | |||
XM_011512428.2 | XP_011510730.1 | 5 | 1318 | |||
XM_011512431.2 | XP_011510733.1 | 4 | 1283 | |||
XM_011512432.2 | XP_011510734.1 | 4 | 1283 | |||
XM_017005711.1 | XP_016861200.1 | 7 | 1502 | |||
XM_017005712.1 | XP_016861201.1 | 6 | 1606 | |||
XM_017005713.1 | XP_016861202.1 | 7 | 1553 | |||
XM_017005714.1 | XP_016861203.1 | 5 | 7658 | |||
XM_017005715.1 | XP_016861204.1 | 6 | 1423 | |||
XM_017005716.1 | XP_016861205.1 | 4 | 1283 | |||
GenBank mRNA | AM774225.1 | - | 3 | 1058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
