siRNA Details
Gene Symbol: PDCD6IPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037599.1 | 10 | 1281 | |||
GenBank mRNA | DB073810.1 | - | 4 | 455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
