siRNA Details
Gene Symbol: MDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027042.1 | 6 | 747 | |||
GenBank mRNA | AJ310434.2 | - | 6 | 747 | ||
BC041472.1 | - | 6 | 1499 | |||
BC069164.1 | - | 6 | 747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
