siRNA Details
Gene Symbol: RBMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002898.3 | NP_002889.1 | 14 | 6924 | ||
XM_005269059.4 | XP_005269116.1 | 14 | 6787 | |||
XM_005269060.4 | XP_005269117.1 | 13 | 6700 | |||
XM_006719541.3 | XP_006719604.1 | 14 | 6649 | |||
XM_006719542.3 | XP_006719605.1 | 13 | 6562 | |||
XM_006719543.3 | XP_006719606.1 | 13 | 6567 | |||
XM_011538637.2 | XP_011536939.1 | 14 | 6593 | |||
XM_011538638.2 | XP_011536940.1 | 15 | 6915 | |||
XM_011538640.2 | XP_011536942.1 | 14 | 6788 | |||
XM_011538642.2 | XP_011536944.1 | 12 | 6379 | |||
XM_017019775.1 | XP_016875264.1 | 14 | 6731 | |||
XM_017019776.1 | XP_016875265.1 | 10 | 6073 | |||
XM_017019777.1 | XP_016875266.1 | 13 | 6497 | |||
XM_017019778.1 | XP_016875267.1 | 10 | 6232 | |||
GenBank mRNA | AK096412.1 | - | 1 | 1226 | ||
AK097342.1 | - | 1 | 1650 | |||
AK125252.1 | - | 1 | 2605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
