siRNA Details
Gene Symbol: ZSCAN5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080456.2 | NP_001073925.2 | 4 | 1106 | ||
XM_005258864.3 | XP_005258921.2 | 5 | 1265 | |||
XM_006723189.3 | XP_006723252.1 | 5 | 1286 | |||
GenBank mRNA | BX094413.1 | - | 3 | 664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
