siRNA Details
Gene Symbol: ZNF766 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010851.2 | NP_001010851.1 | 4 | 2707 | ||
XM_011527460.1 | XP_011525762.1 | 4 | 3034 | |||
XM_017027441.1 | XP_016882930.1 | 2 | 2771 | |||
GenBank mRNA | AK024268.1 | - | 1 | 2344 | ||
AK097729.1 | - | 6 | 2064 | |||
BX538183.1 | - | 4 | 2679 | |||
BX648899.1 | - | 4 | 2707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
