siRNA Details
Gene Symbol: LOC105372315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528468.2 | XP_011526770.1 | 2 | 2439 | ||
XM_011528469.2 | XP_011526771.1 | 2 | 1839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
