siRNA Details
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003262.3 | NP_003253.1 | 8 | 6175 | ||
GenBank mRNA | AK057365.1 | - | 1 | 1538 | ||
BC089399.1 | - | 1 | 1537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
