siRNA Details
Gene Symbol: TSPAN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080509.2 | NP_001073978.1 | 6 | 582 | ||
XM_011520679.2 | XP_011518981.1 | 6 | 844 | |||
XM_011520680.2 | XP_011518982.1 | 6 | 822 | |||
GenBank mRNA | BC127661.1 | - | 4 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
