siRNA Details
Gene Symbol: FAM230C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027278.1 | 8 | 1341 | |||
GenBank mRNA | AK097777.1 | - | 8 | 1281 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
