siRNA Details
Gene Symbol: PABPC1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026904.1 | 1 | 1704 | |||
GenBank mRNA | AF132027.1 | - | 1 | 1335 | ||
AK292357.1 | - | 1 | 1317 | |||
BC033032.1 | - | 1 | 1319 | |||
BC068242.1 | - | 1 | 1068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
