siRNA Details
Gene Symbol: PABPC1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026904.1 | 1 | 2422 | |||
GenBank mRNA | BC033032.1 | - | 1 | 2037 | ||
BC068242.1 | - | 1 | 1786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
