siRNA Details
Gene Symbol: UBA52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033930.2 | NP_001029102.1 | 5 | 1481 | ||
NM_001321017.1 | NP_001307946.1 | 5 | 1480 | |||
NM_001321018.1 | NP_001307947.1 | 5 | 2107 | |||
NM_001321019.1 | NP_001307948.1 | 5 | 1645 | |||
NM_001321020.1 | NP_001307949.1 | 5 | 1679 | |||
NM_001321021.1 | NP_001307950.1 | 5 | 1494 | |||
NM_001321022.1 | NP_001307951.1 | 4 | 1394 | |||
NM_003333.4 | NP_003324.1 | 5 | 1459 | |||
GenBank mRNA | AF545864.1 | - | 1 | 686 | ||
AK057790.1 | - | 1 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
