siRNA Details
Gene Symbol: LINC00638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024396.1 | 1 | 1435 | |||
GenBank mRNA | AK095404.1 | - | 1 | 1435 | ||
AK292179.1 | - | 1 | 699 | |||
BC015579.2 | - | 1 | 697 | |||
BC041036.1 | - | 1 | 697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
