siRNA Details
Gene Symbol: LOC100133315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029192.1 | 3, 4 | 430 | |||
GenBank mRNA | BX102226.1 | - | 2, 3 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
