siRNA Details
Gene Symbol: ADGRE4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024075.1 | 3 | 229 | |||
GenBank mRNA | AF489700.1 | - | 3 | 229 | ||
AY181245.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
