siRNA Details
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 11 | 5761 | ||
NM_080616.4 | NP_542183.2 | 8 | 5100 | |||
GenBank mRNA | AL122043.2 | - | 1 | 3366 | ||
AL834520.1 | - | 1 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
