siRNA Details
Gene Symbol: DKFZP434A062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026964.3 | 1 | 2370 | |||
GenBank mRNA | AK127730.1 | - | 1 | 2370 | ||
AL080191.1 | - | 1 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
