siRNA Details
Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 11 | 1214 | |||
GenBank mRNA | AJ132821.1 | - | 10 | 1148 | ||
AJ132822.1 | - | 10 | 1030 | |||
AJ132823.1 | - | 9 | 964 | |||
AJ132826.1 | - | 1 | 144 | |||
AJ132827.1 | - | 1 | 144 | |||
BC047448.1 | - | 11 | 1348 | |||
BC067864.1 | - | 9 | 1007 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
