siRNA Details
Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 12 | 1365 | |||
GenBank mRNA | AJ132821.1 | - | 11 | 1299 | ||
AJ132822.1 | - | 11 | 1181 | |||
AJ132823.1 | - | 10 | 1115 | |||
AJ132827.1 | - | 2 | 295 | |||
BC047448.1 | - | 12 | 1499 | |||
BC067864.1 | - | 10 | 1158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
