siRNA Details
Gene Symbol: SIRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122962.1 | NP_001116434.1 | 5 | 2503 | ||
NM_001134836.1 | NP_001128308.1 | 5 | 2209 | |||
GenBank mRNA | AK095499.1 | - | 5 | 2308 | ||
BG539998.1 | - | 1 | 663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
