siRNA Details
Gene Symbol: PLXNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020911.1 | NP_065962.1 | 32 | 8319 | ||
XM_005250686.4 | XP_005250743.1 | 32 | 8717 | |||
XM_006716171.3 | XP_006716234.1 | 33 | 8810 | |||
XM_017012779.1 | XP_016868268.1 | 31 | 8024 | |||
GenBank mRNA | AB046770.3 | - | 30 | 6784 | ||
AK057743.1 | - | 1 | 1275 | |||
AK091538.1 | - | 4 | 3073 | |||
AK095606.1 | - | 8 | 3552 | |||
AL834504.1 | - | 7 | 3424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
