siRNA Details
Gene Symbol: LINC01106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027244.1 | 2 | 1033 | |||
GenBank mRNA | AK095678.1 | - | 2 | 1342 | ||
CR749351.1 | - | 3 | 1124 |
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 4 | 2449 | |||
NR_046111.1 | 3 | 2264 | ||||
NR_046112.1 | 2 | 1022 | ||||
GenBank mRNA | AK056084.1 | - | 4 | 2366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
