siRNA Details
Gene Symbol: ATG2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720187.2 | XP_006720250.1 | 12 | 2123 | ||
XM_017021397.1 | XP_016876886.1 | 12 | 2126 | |||
GenBank mRNA | AK055200.1 | - | 12 | 2126 | ||
BU167202.1 | - | 1 | 16 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
